Please login to your online account to display your discounted pricing

IgG2a Rat anti-Mouse, APC, Clone: m2a-15F8, eBioscience™

Rat Monoclonal Antibody

$114.55 - $245.05

Specifications

Antigen IgG2a
Clone m2a-15F8
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123127

 
affymetrix
17421080
25μg Each for $114.55

501129147

 
affymetrix
17421082
100μg Each for $245.05
Description & Specifications

Specifications

Antigen IgG2a
Clone m2a-15F8
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody m2a-15F8 recognizes Mouse IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG2a or any rat isotype antibodies.