Please login to your online account to display your discounted pricing

IgG2a Mouse anti-Rat, PE-Cy7, Clone: r2a-21B2, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $288.55

Specifications

Antigen IgG2a
Clone r2a-21B2
Host Species Murine
Species Reactivity Rat
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123373

 
life technologies
LS25481780
25μg Each for $114.55

501123374

 
life technologies
LS25481782
100μg Each for $288.55
Description & Specifications

Specifications

Antigen IgG2a
Clone r2a-21B2
Host Species Murine
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody r2a-21B2 recognizes Rat IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2a or any mouse isotype antibodies.