missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2a Mouse anti-Rat, eFluor 660, Clone: r2a-21B2, eBioscience™

Mouse Monoclonal Antibody

$91.08 - $223.08

Specifications

Antigen IgG2a
Clone r2a-21B2
Host Species Murine
Species Reactivity Rat
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124372 Life Technologies
LS50481780
25μg Each for $91.08
501124373 Life Technologies
LS50481782
100μg Each for $223.08
Specifications

Description

The monoclonal antibody r2a-21B2 recognizes Rat IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2a or any mouse isotype antibodies.
Description & Specifications

Specifications

IgG2a
Murine
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
r2a-21B2
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer