missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2a, K Rat, eFluor 570, Clone: eBR2a, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$146.52 - $361.68

Specifications

Antigen IgG2a, K
Clone eBR2a
Host Species Rat
Applications Immunocytochemistry,Immunohistochemistry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123613 Life Technologies
LS41432180
25μg Each for $146.52
501123614 Life Technologies
LS41432182
100μg Each for $361.68
Specifications

Description

The monoclonal rat IgG2a is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG2a, K
Rat
RUO
Conjugated
Monoclonal
0.2mg/mL
eBR2a
Immunocytochemistry,Immunohistochemistry
eFluor 570
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer