Learn More
Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 570, eBioscience™, Invitrogen™
Rat Isotype Control
Supplier: Invitrogen 41432180
Description
Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin. Applications Reported: This eBR2a antibody has been reported for use in immunohistochemical and immunocytochemical staining. Applications Tested: This Rat IgG2a Isotype Control eFluor® 570 has been tested by immunocytochemistry on fixed and permeabilized C2C12 cells and by immunohistochemistry (IHC-P) on formalin-fixed paraffin embedded human tissue. Use isotype control at the same concentration as experimental antibody. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin.Applications Reported: This eBR2a antibody has been reported for use in immunohistochemical and immunocytochemical staining.Applications Tested: This Rat IgG2a Isotype Control eFluor™ 570 has been tested by immunocytochemistry on fixed and permeabilized C2C12 cells and by immunohistochemistry (IHC-P) on formalin-fixed paraffin embedded human tissue. Use isotype control at the same concentration as experimental antibody. Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable.Excitation: 555 nm; Emission: 570 nmSpecifications
Rat IgG2a kappa | |
eBR2a | |
eFluor 570 | |
IgG2, IgG2a, IgG2k | |
Affinity chromatography | |
RUO | |
None | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Rat | |
25 μg | |
Isotype Control (Primary) | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
IgG2a κ |
For Research Use Only.