missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2a, K Rat, Clone: eBR2a, Isotype Control, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$77.88 - $236.28

Specifications

Antigen IgG2a, K
Clone eBR2a
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122958 Life Technologies
LS16432181
50μg Each for $77.88
5014529 Life Technologies
LS16432182
100μg Each for $130.68
501129674 Life Technologies
LS16432185
500μg Each for $236.28
Specifications

Description

The monoclonal rat IgG2a, κ is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG2a, K
Rat
RUO
Purified
Monoclonal
1mg/mL
eBR2a
Flow Cytometry
Unlabeled
Store at 2-8°C.
aqueous buffer, no sodium azide