Please login to your online account to display your discounted pricing

IgG2a, K Mouse, PE, Clone: eBM2a, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$85.55 - $201.55

Specifications

Antigen IgG2a, K
Clone eBM2a
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122314

 
life technologies
LS12472482
100μg Each for $201.55

501128747

 
life technologies
LS12472442
100 tests Pack for $172.55

501129434

 
life technologies
LS12472481
50μg Each for $129.05

501129452

 
life technologies
LS12472441
25 tests Each for $85.55
Description & Specifications

Specifications

Antigen IgG2a, K
Clone eBM2a
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.5μg)/test

This is a monoclonal mouse IgG2a, κ antibody. It is used as an isotype control for mouse IgG2a antibodies.