missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2a, K Mouse, eFluor 450, Clone: eBM2a, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$143.55 - $288.55

Specifications

Antigen IgG2a, K
Clone eBM2a
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016354 Life Technologies
LS48472482
100μg Each for $288.55
501128790 Life Technologies
LS48472480
25μg Each for $143.55
Specifications

Description

This is a monoclonal mouse IgG2a, κ antibody. It is used as an isotype control for mouse IgG2a antibodies.
Description & Specifications

Specifications

IgG2a, K
Murine
RUO
Conjugated
Monoclonal
0.2mg/mL
eBM2a
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer