missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Isotype Control
Supplier: Thermo Fisher Scientific 48472482
Description
Description: This is a monoclonal mouse IgG2a, kappa antibody. It is used as an isotype control for mouse IgG2a antibodies. Applications Reported: Mouse IgG2a K Isotype Control eFluor® 450 has been reported for use in flow cytometric analysis. Applications Tested: eFluor® 450 Mouse IgG2a, K Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2a kappa Control antibody specifically detects Mouse IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG2a kappa | |
eBM2a | |
eFluor 450 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2a κ |
For Research Use Only.