Please login to your online account to display your discounted pricing

IgG2a, K Mouse, Clone: eBM2a, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$143.55 - $288.55

Specifications

Antigen IgG2a, K
Clone eBM2a
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124963

 
life technologies
LS69472480
25μg Each for $143.55

501124964

 
life technologies
LS69472482
100μg Each for $288.55

501129647

 
life technologies
LS14472485
500μg Each for $187.05
Description & Specifications

Specifications

Antigen IgG2a, K
Clone eBM2a
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This is a monoclonal mouse IgG2a antibody. It is used as an isotype control for mouse IgG2a antibodies.