missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG1 Rat anti-Mouse, HRP, Clone: M1-14D12, eBioscience™

Rat Monoclonal Antibody

$77.88 - $196.68

Specifications

Antigen IgG1
Clone M1-14D12
Host Species Rat
Applications ELISA,Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Western Blot
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123247 Life Technologies
LS18401580
25μg Each for $77.88
501128837 Life Technologies
LS18401582
100μg Each for $196.68
Specifications

Description

The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies.
Description & Specifications

Specifications

IgG1
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
M1-14D12
ELISA,Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Western Blot
HRP
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL