Please login to your online account to display your discounted pricing

IgG1 Rat anti-Mouse, eFluor 615, Clone: M1-14D12, eBioscience™

Rat Monoclonal Antibody

$131.95 - $323.35

Specifications

Antigen IgG1
Clone M1-14D12
Host Species Rat
Species Reactivity Mouse
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123668

 
life technologies
LS42401580
25μg Each for $131.95

501123669

 
life technologies
LS42401582
100μg Each for $323.35
Description & Specifications

Specifications

Antigen IgG1
Clone M1-14D12
Host Species Rat
Species Reactivity Mouse
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate eFluor 615
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies.