missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG1 Rat anti-Mouse, Biotin, Clone: M1-14D12, eBioscience™

Rat Monoclonal Antibody

$79.20 - $200.64

Specifications

Antigen IgG1
Clone M1-14D12
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry,Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122540 Life Technologies
LS13401582
100μg Each for $200.64
501129145 Life Technologies
LS13401580
25μg Each for $79.20
Specifications

Description

The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy applications. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies.
Description & Specifications

Specifications

IgG1
Rat
Flow Cytometry,Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen)
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
M1-14D12
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer