Please login to your online account to display your discounted pricing

IgG1 Mouse anti-Rat, PE, Clone: R1-12D10, eBioscience™

Mouse Monoclonal Antibody

$100.05 - $245.05

Specifications

Antigen IgG1
Clone R1-12D10
Host Species Mouse
Species Reactivity Rat
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122315

 
affymetrix
12481280
25μg Each for $100.05

501129274

 
affymetrix
12481282
100μg Each for $245.05
Description & Specifications

Specifications

Antigen IgG1
Clone R1-12D10
Host Species Mouse
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The R1-12D10 monoclonal antibody recognizes rat IgG1 antibodies and can be used as a second step reagent in microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG1.