missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG1 Mouse anti-Rat, FITC, Clone: R1-12D10, eBioscience™

Mouse Monoclonal Antibody

$106.92 - $200.64

Specifications

Antigen IgG1
Clone R1-12D10
Host Species Murine
Applications Flow Cytometry,Immunocytochemistry,Immunohistochemistry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122128 Life Technologies
LS11481280
25μg Each for $106.92
501122129 Life Technologies
LS11481282
100μg Each for $200.64
Specifications

Description

The R1-12D10 monoclonal antibody recognizes rat IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG1.
Description & Specifications

Specifications

IgG1
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
R1-12D10
Flow Cytometry,Immunocytochemistry,Immunohistochemistry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL