Please login to your online account to display your discounted pricing

IgG1, K Rat, PE-eFluor 610, Clone: eBRG1, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$100.05 - $245.05

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124811

 
life technologies
LS61430180
25μg Each for $100.05

501124812

 
life technologies
LS61430182
100μg Each for $245.05
Description & Specifications

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.