Please login to your online account to display your discounted pricing

IgG1, K Rat, PE-Cy5, Clone: eBRG1, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$230.55 - $317.55

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014106

 
life technologies
LS15430181
50μg Each for $230.55

5014107

 
life technologies
LS15430182
100μg Each for $317.55
Description & Specifications

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.