missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG1, K Rat, PE, Clone: eBRG1, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$133.40 - $332.05

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5018641 Life Technologies
LS12430183
200μg Each for $332.05
501129615 Life Technologies
LS12430182
100μg Each for $220.40
501129619 Life Technologies
LS12430181
50μg Each for $133.40
Specifications

Description

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG1, K
Rat
RUO
Conjugated
Monoclonal
0.2mg/mL
eBRG1
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer