missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG1, K Rat, FITC, Clone: eBRG1, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$105.60 - $275.88

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry,Immunocytochemistry,Immunohistochemistry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122121 Life Technologies
LS11430181
50μg Each for $105.60
5018602 Life Technologies
LS11430182
100μg Each for $187.44
5018603 Life Technologies
LS11430185
500μg Each for $275.88
Specifications

Description

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG1, K
Rat
RUO
Conjugated
Monoclonal
0.5mg/mL
eBRG1
Flow Cytometry,Immunocytochemistry,Immunohistochemistry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer