Learn More
Rat IgG1 kappa, eFluor 660, clone: eBRG1, Isotype Control, eBioscience™
Rat Isotype Control
Manufacturer: Life Technologies LS50430182
Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: eFluor™ 660 Rat IgG1 Isotype Control has been reported for use in flow cytometric analysis and immunohistochemical staining.Applications Tested: eFluor™ 660 Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse spleen cells and normal human peripheral blood lymphocytes. Isotype control should be used at the same concentration as experimental antibody.eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
Rat IgG1 kappa | |
Control | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
IgG1, IgG1k | |
IgG1, kappa, kappa | |
100μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Monoclonal |
Control, Flow Cytometry, Immunohistochemistry | |
eBRG1 | |
eFluor 660 | |
Liquid | |
Rat | |
Affinity chromatography | |
RUO | |
Isotype Control | |
Not Applicable |
For Research Use Only.