Please login to your online account to display your discounted pricing

IgG1, K Rat, eFluor 570, Clone: eBRG1, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$160.95 - $397.30

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Immunocytochemistry
Applications Immunohistochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123609

 
life technologies
LS41430180
25μg Each for $160.95

501123610

 
life technologies
LS41430182
100μg Each for $397.30
Description & Specifications

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Immunocytochemistry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.