missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG1, K Rat, eFluor 450, Clone: eBRG1, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$104.28 - $236.28

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124162 Life Technologies
LS48430182
100μg Each for $236.28
5016346 Life Technologies
LS48430180
25μg Each for $104.28
Specifications

Description

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG1, K
Rat
RUO
Conjugated
Monoclonal
0.2mg/mL
eBRG1
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer