missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG1, K Rat, Clone: eBRG1, Isotype Control, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$77.88 - $236.28

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014526 Life Technologies
LS16430181
50μg Each for $77.88
5014527 Life Technologies
LS16430185
500μg Each for $236.28
Specifications

Description

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG1, K
Rat
RUO
Purified
Monoclonal
1mg/mL
eBRG1
Flow Cytometry
Unlabeled
Store at 2-8°C.
aqueous buffer, no sodium azide