missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG1, K Rat, Clone: eBRG1, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$64.68 - $157.08

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry,Immunocytochemistry,Immunohistochemistry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013034 Life Technologies
LS14430181
50μg Each for $64.68
5013035 Life Technologies
LS14430182
100μg Each for $104.28
5013036 Life Technologies
LS14430185
500μg Each for $157.08
Specifications

Description

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG1, K
Rat
RUO
Purified
Monoclonal
0.5mg/mL
eBRG1
Flow Cytometry,Immunocytochemistry,Immunohistochemistry
Unlabeled
Store at 2-8°C.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer