Please login to your online account to display your discounted pricing

IgG1, K Rat, Biotin, Clone: eBRG1, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$85.55 - $390.05

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011852

 
life technologies
LS13430181
50μg Each for $85.55

5011853

 
life technologies
LS13430182
100μg Each for $158.05

5011854

 
life technologies
LS13430185
500μg Each for $390.05
Description & Specifications

Specifications

Antigen IgG1, K
Clone eBRG1
Host Species Rat
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal rat IgG1, κ is useful as an isotype control immunoglobulin.