missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rat IgG1 kappa Isotype Control (eBRG1), APC, eBioscience™, Invitrogen™
Rat Isotype Control
$252.00 - $542.00
Specifications
Antigen | Rat IgG1 kappa |
---|---|
Clone | eBRG1 |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | APC |
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG1 isotype control has been reported for use in flow cytometric analysis and ELISA. Applications Tested: This rat IgG1 isotype control has been used by flow cytometric analysis of mouse splenocyte suspension, This can be used at the same concentration as the experimental antibody. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG1 kappa Control antibody specifically detects Rat IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG1 kappa | |
0.2 mg/mL | |
APC | |
Rat | |
Not Applicable | |
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBRG1 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.