Please login to your online account to display your discounted pricing

IgG1, K Mouse, PE-Cy5.5, Clone: P3.6.2.8.1, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$114.55 - $288.55

Specifications

Antigen IgG1, K
Clone P3.6.2.8.1
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123553

 
affymetrix
35471480
25μg Each for $114.55

501128769

 
affymetrix
35471482
100μg Each for $288.55
Description & Specifications

Specifications

Antigen IgG1, K
Clone P3.6.2.8.1
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5.5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal mouse IgG1, κ immunoglobulin is useful as an isotype control.