Please login to your online account to display your discounted pricing

IgG1, K Mouse, eFluor 570, Clone: P3.6.2.8.1, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$146.45 - $279.85

Specifications

Antigen IgG1, K
Clone P3.6.2.8.1
Host Species Murine
Applications Immunocytochemistry
Applications Immunohistochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123616

 
life technologies
LS41471480
25μg Each for $146.45

501123617

 
life technologies
LS41471482
100μg Each for $279.85
Description & Specifications

Specifications

Antigen IgG1, K
Clone P3.6.2.8.1
Host Species Murine
Applications Immunocytochemistry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal mouse IgG1, κ immunoglobulin is useful as an isotype control.