Please login to your online account to display your discounted pricing

IgG1, K Mouse, Clone: P3.6.2.8.1, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$71.05 - $71.05

Specifications

Antigen IgG1, K
Clone P3.6.2.8.1
Host Species Murine
Applications Flow Cytometry
Applications Flow Cytometry (Intracellular Staining)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013045

 
life technologies
LS14471481
50μg Each for $71.05
Description & Specifications

Specifications

Antigen IgG1, K
Clone P3.6.2.8.1
Host Species Murine
Applications Flow Cytometry
Applications Flow Cytometry (Intracellular Staining)
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal mouse IgG1, κ immunoglobulin is useful as an isotype control.