Please login to your online account to display your discounted pricing

IgG, Whole Molec. PAb, anti-Sheep, Rabbit, Liq., MP Biomedicals

Manufacturer: mp biomedicals  55802

 View all options for this product

Catalog No. ICN55802


Description & Specifications


Antigen IgG, Whole Molecule
Conjugate Unlabeled
Host Species Rabbit
Isotype IgG
Quantity 5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal