missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG Golden Syrian Hamster, PE-eFluor 610, Polyclonal, Isotype Control, eBioscience™

Golden Syrian Hamster Polyclonal Isotype Control

$130.68 - $328.68

Specifications

Antigen IgG
Host Species Golden Syrian Hamster
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124829 Life Technologies
LS61491480
25μg Each for $130.68
501124830 Life Technologies
LS61491482
100μg Each for $328.68
Specifications

Description

This hamster IgG fraction was purified by affinity chromatography from normal Golden Syrian hamster serum.
Description & Specifications

Specifications

IgG
Flow Cytometry
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Golden Syrian Hamster
RUO
Affinity Purified
Polyclonal
0.2mg/mL