Please login to your online account to display your discounted pricing

IgG Fc, γ-Specific PAb, anti-Rabbit, Goat, Liq., MP Biomedicals

Manufacturer: mp biomedicals  55625

 View all options for this product

Catalog No. ICN55625


Description & Specifications


Antigen IgG Fc, gamma-Specific
Conjugate Unlabeled
Host Species Goat
Isotype IgG
Quantity 5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal