Please login to your online account to display your discounted pricing

IgG Fc, γ-Specific PAb, anti-Rabbit, Goat, Liq., MP Biomedicals

$261.35 - $261.35

Specifications

Antigen IgG Fc, gamma-Specific
Host Species Goat
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

ICN55625

 
mp biomedicals
55625
Unlabeled 5mL Each for $261.35
Description & Specifications

Specifications

Antigen IgG Fc, gamma-Specific
Host Species Goat
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly