Please login to your online account to display your discounted pricing

IgG, F(ab')2 Goat anti-Mouse, PerCP-eFluor 710, Polyclonal, eBioscience™

Goat Polyclonal Antibody

$210.98 - $249.40

Specifications

Antigen IgG, F(ab')2
Host Species Goat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501128763

 
affymetrix
46401082
100μg Each for $249.40

501130806

 
affymetrix
46401080
25μg Each for $210.98
Description & Specifications

Specifications

Antigen IgG, F(ab')2
Host Species Goat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The goat anti-mouse IgG polyclonal antibody reacts with the heavy and light chains of mouse IgG, with minimal cross reactivity to human, rat, bovine, goat, hamster, and rabbit serum proteins. Reactivity has been shown to IgG1, IgG2a, IgG2b, and IgG3, as well as some reactivity to IgM. The antibody is a F(ab')2 fragment which contains the 2 Fab domains still linked through the hinge region of the immunoglobulin. Removal of the Fc region helps to reduce issues typically found with Fc-mediated binding.

This polyclonal has been validated as a secondary reagent (its ability to recognize cells stained with a mouse IgG followed by this polyclonal antibody).