Please login to your online account to display your discounted pricing

IgG Armenian Hamster, PE-Cy7, Clone: eBio299Arm, Isotype Control, eBioscience™

Armenian Hamster Monoclonal Isotype Control

$205.90 - $404.55

Specifications

Antigen IgG
Clone eBio299Arm
Host Species Armenian Hamster
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015574

 
life technologies
LS25488881
50μg Each for $205.90

5015575

 
life technologies
LS25488882
100μg Each for $404.55
Description & Specifications

Specifications

Antigen IgG
Clone eBio299Arm
Host Species Armenian Hamster
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.