missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG Armenian Hamster, PE-Cy5, Clone: eBio299Arm, Isotype Control, eBioscience™

Armenian Hamster Monoclonal Isotype Control

$157.08 - $262.68

Specifications

Antigen IgG
Clone eBio299Arm
Host Species Armenian Hamster
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014128 Life Technologies
LS15488881
50μg Each for $157.08
5014129 Life Technologies
LS15488882
100μg Each for $262.68
Specifications

Description

The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG
Armenian Hamster
RUO
Conjugated
Monoclonal
0.2mg/mL
eBio299Arm
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer