Please login to your online account to display your discounted pricing

IgG Armenian Hamster, Clone: eBio299Arm, Isotype Control, Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody, Functional Grade

$100.05 - $288.55

Specifications

Antigen IgG
Clone eBio299Arm
Host Species Armenian Hamster
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014545

 
life technologies
LS16488881
50μg Each for $100.05

501129592

 
life technologies
LS16488885
500μg Each for $288.55
Description & Specifications

Specifications

Antigen IgG
Clone eBio299Arm
Host Species Armenian Hamster
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.