Please login to your online account to display your discounted pricing

IgG Armenian Hamster, Clone: eBio299Arm, Isotype Control, eBioscience™

Armenian Hamster Monoclonal Isotype Control

$114.55 - $259.55

Specifications

Antigen IgG
Clone eBio299Arm
Host Species Armenian Hamster
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124967

 
life technologies
LS69488880
25μg Each for $114.55

501124968

 
life technologies
LS69488882
100μg Each for $259.55

501129491

 
life technologies
LS14488885
500μg Each for $187.05
Description & Specifications

Specifications

Antigen IgG
Clone eBio299Arm
Host Species Armenian Hamster
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 506
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.