Learn More
Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 506, eBioscience™, Invitrogen™
Armenian Hamster Isotype Control
Supplier: Invitrogen 69488880
Description
Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody. eFluor® 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody.eFluor™ 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
Armenian Hamster IgG | |
eBio299Arm | |
eFluor 506 | |
IgG | |
Affinity chromatography | |
RUO | |
None | |
Liquid |
Flow Cytometry | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Armenian Hamster | |
25 μg | |
Isotype Control (Primary) | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
IgG |
For Research Use Only.