missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Armenian Hamster IgG Isotype Control (eBio299Arm), Biotin, eBioscience™, Invitrogen™

Armenian Hamster Isotype Control

$129.00 - $411.00

Specifications

Antigen Armenian Hamster IgG
Clone eBio299Arm
Concentration 0.5 mg/mL
Applications Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry
Conjugate Biotin
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011876
View Documents Promotion Details
Invitrogen
13488881
50 μg
Each for $129.00
 
5011877
View Documents Promotion Details
Invitrogen
13488885
500 μg
Each for $411.00
 
Description

Description

Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood and can be used at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Specifications

Specifications

Armenian Hamster IgG
0.5 mg/mL
Biotin
Armenian Hamster
Not Applicable
IgG; Immunoglobulin G; ImmunoglobulinG
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
eBio299Arm
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
IgG
Affinity chromatography
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only.