Learn More
Armenian Hamster IgG, APC, clone: eBio299Arm, Isotype Control, eBioscience™
Armenian Hamster Isotype Control
Manufacturer: Life Technologies LS17488881
Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis. Isotype control should be used at the same concentration as experimental antibody.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
Armenian Hamster IgG | |
Control | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
IgG | |
IgG | |
50μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Monoclonal |
Control, Flow Cytometry | |
eBio299Arm | |
APC | |
Liquid | |
Armenian Hamster | |
Affinity chromatography | |
RUO | |
Isotype Control | |
Not Applicable |
For Research Use Only.