missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ IGF-I Ab-1, Mouse Monoclonal Antibody

Ideal for Western Blotting, Thermo Scientific™ IGF-I Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$276.86 - $1,408.78

Specifications

Antigen IGF-I Ab-1
Clone M23
Applications Western Blot
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1508P Thermo Scientific™ Lab Vision™
MS1508P
200μg/mL 1mL Each for $1,282.55
MS1508P0 Thermo Scientific™ Lab Vision™
MS1508P0
200μg/mL 100μL Each for $276.86
MS1508P1 Thermo Scientific™ Lab Vision™
MS1508P1
200μg/mL 500μL Each for $776.16
MS1508P1BX Thermo Scientific™ Lab Vision™
MS1508P1BX
1mg/mL 100μL Each for $858.86
MS1508PBX Thermo Scientific™ Lab Vision™
MS1508PBX
1mg/mL 200μL Each for $1,408.78
Specifications

Description

Ideal for Western Blotting, Thermo Scientific™ IGF-I Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insulin-like growth factor 1 (IGF-1) is involved in regulation of neuronal growth and development in central and peripheral nervous system. It is known to protect neurons against cell death induced by amyloidogenic derivatives, glucose or serum deprivation through pathways involving AKT kinase and transcription factor FKHRL1 phosphorylation. Activation of the insulin-like growth factor system has emerged as a key factor for tumor progression and resistance to apoptosis in many cancers like breast and thyroid cancers.

Host Species: Mouse

Clone: M23

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Immunogen: Purified human IGF-1

Molecular Weight: ∽7.6kDa

Positive Control: Pancreas, IGF-1 recombinant protein

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

IGF-I Ab-1
Western Blot
Unlabeled
Murine
Growth Factor
Purified human IGF-1
M23
Monoclonal
Human,Mouse,Rat,Human,Mouse,Rat,Human,Mouse,Rat
RUO
Primary
IgG1