Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ IGF-I Ab-1, Mouse Monoclonal Antibody


Ideal for Western Blotting, Thermo Scientific™ IGF-I Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1508P1BX

 View more versions of this product

Catalog No. MS1508P1BX

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Immunogen Purified human IGF-1
Regulatory Status RUO
Host Species Mouse
Isotype IgG1
Clone M23
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen IGF-I Ab-1
Format Purified
Quantity 100µL
Type Primary
Conjugate Unlabeled
Research Discipline Growth Factor
Classification Monoclonal
Concentration 1mg/mL

Ideal for Western Blotting, Thermo Scientific™ IGF-I Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insulin-like growth factor 1 (IGF-1) is involved in regulation of neuronal growth and development in central and peripheral nervous system. It is known to protect neurons against cell death induced by amyloidogenic derivatives, glucose or serum deprivation through pathways involving AKT kinase and transcription factor FKHRL1 phosphorylation. Activation of the insulin-like growth factor system has emerged as a key factor for tumor progression and resistance to apoptosis in many cancers like breast and thyroid cancers.

Host Species: Mouse

Clone: M23

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Immunogen: Purified human IGF-1

Molecular Weight: ∽7.6kDa

Positive Control: Pancreas, IGF-1 recombinant protein

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO