Please login to your online account to display your discounted pricing

Thermo Scientific™ IGF-1R (β -Subunit) Ab-5, Mouse Monoclonal Antibody

Ideal for a variety of molecular biology techniques, Thermo Scientific™ IGF-1R (β-Subunit) Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results. 

$280.98 - $1,417.24

Specifications

Antigen IGF-1R (beta-Subunit) Ab-5
Clone 1-2
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS645P

 
thermo scientific™
MS645P
200μg/mL 1mL Each for $1,315.71

MS645P0

 
thermo scientific™
MS645P0
200μg/mL 100µL Each for $280.98

MS645P1

 
thermo scientific™
MS645P1
200μg/mL 500µL Each for $808.27

MS645P1ABX

 
thermo scientific™
MS645P1ABX
1mg/mL 100µL Each for $909.67

MS645PABX

 
thermo scientific™
MS645PABX
1mg/mL 200µL Each for $1,417.24
Description & Specifications

Specifications

Antigen IGF-1R (beta-Subunit) Ab-5
Clone 1-2
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen A 15-mer peptide corresponding to aa 1323-1337 (Tyr-RKNERALPLPQSSTC) from the C-terminal of Beta-subunit of human IGF-1R
Isotype IgG2b kappa

Ideal for a variety of molecular biology techniques, Thermo Scientific™ IGF-1R (β-Subunit) Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results. 

IGF-1R consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.

Host Species: Mouse

Clone: 1-2

Isotype: IgG2b/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1323-1337

Immunogen: A 15-mer peptide corresponding to aa 1323-1337 (Tyr-RKNERALPLPQSSTC) from the C-terminal of β-subunit of human IGF-1R

Molecular Weight: 95kDa

Positive Control: Placenta or Breast CA

Cellular Localization: Membrane Ab-5 is the antibody-of-choice for Western blotting of Beta-subunit of IGF-1R. Ab-3 reacts only weakly with hybrids of Insulin Receptor and IGF-1R while Ab-5 reacts very well. So, Ab-3, in conjunction with Ab-5, has proved useful in the isolation of hybrid receptors.

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Not optimum)
  • Affinity Purification of HYBRID and IGF-1R

Note:

USA: RUO; Int'l: RUO