Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ IGF-1R (α -Subunit) Ab-4, Mouse Monoclonal Antibody

Ideal for immunoprecipitation, Thermo Scientific™ IGF-1R (α-Subunit) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

$273.43 - $1,373.92

Specifications

Antigen IGF-1R (alpha-Subunit) Ab-4
Clone 24-60
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS644P

 
thermo scientific™ lab vision™
MS644P
200μg/mL 1mL Each for $1,265.04

MS644P0

 
thermo scientific™ lab vision™
MS644P0
200μg/mL 100μL Each for $273.43

MS644P1

 
thermo scientific™ lab vision™
MS644P1
200μg/mL 500μL Each for $795.12

MS644P1ABX

 
thermo scientific™ lab vision™
MS644P1ABX
1mg/mL 100μL Each for $862.04

MS644PABX

 
thermo scientific™ lab vision™
MS644PABX
1mg/mL 200μL Each for $1,373.92
Description & Specifications

Specifications

Antigen IGF-1R (alpha-Subunit) Ab-4
Clone 24-60
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Isotype IgG2a kappa

Ideal for immunoprecipitation, Thermo Scientific™ IGF-1R (α-Subunit) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.

Host Species: Mouse

Clone: 24-60

Isotype: IgG2a/κ

Species Reactivity: Human. Weakly reacts with Rabbit and Pig (Ab-2 is better). Does not react with rat.

Epitope: aa 184-283

Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA

Molecular Weight: 125kDa

Positive Control: Placenta or Breast CA

Cellular Localization: Membrane Ab-4 is excellent for immunoprecipitation of alpha-subunit of IGF-1R.

Recommended for:

  • Western Blotting (Not suitable)
  • Immunoprecipitation
  • Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
  • Substantially (>80%) Inhibits IGF-1 Binding

Note:

USA: RUO; Int'l: RUO