Please login to your online account to display your discounted pricing

Thermo Scientific™ IGF-1 R (α -Subunit) Ab-2, Mouse Monoclonal Antibody

Recommended for Immunoprecipitation, Thermo Scientific™ IGF-1 R (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

$273.86 - $1,377.39

Specifications

Antigen IGF-1R (alpha-Subunit) Ab-2
Clone 17-69
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS642P

 
thermo scientific™
MS642P
200μg/mL 1mL Each for $1,299.88

MS642P0

 
thermo scientific™
MS642P0
200μg/mL 100µL Each for $273.86

MS642P1

 
thermo scientific™
MS642P1
200μg/mL 500µL Each for $805.82

MS642P1ABX

 
thermo scientific™
MS642P1ABX
1mg/mL 100µL Each for $877.06

MS642PABX

 
thermo scientific™
MS642PABX
1mg/mL 200µL Each for $1,377.39
Description & Specifications

Specifications

Antigen IGF-1R (alpha-Subunit) Ab-2
Clone 17-69
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Isotype IgG1

Recommended for Immunoprecipitation, Thermo Scientific™ IGF-1 R (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.

Host Species: Mouse

Clone: 17-69

Isotype: IgG1

Species Reactivity: Human, Pig and Rabbit. Does not react with rat. Others not known.

Epitope: aa 514-586 (exon 7/8)

Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA

Molecular Weight: 125kDa

Positive Control: Placenta or Breast CA

Cellular Localization: Membrane Ab-2 is suitable for studies on pig and rabbit IGF-1R.

Recommended for:

  • Immunoprecipitation
  • Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
  • Does Not Significantly Inhibit IGF Binding

Note:

USA: RUO; Int'l: RUO