Please login to your online account to display your discounted pricing

Thermo Scientific™ IGF-1 R (α -Subunit) Ab-2, Mouse Monoclonal Antibody


Recommended for Immunoprecipitation, Thermo Scientific™ IGF-1 R (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™  MS642P1ABX

 View more versions of this product

Catalog No. MS642P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Format Concentrated
Antigen IGF-1R (alpha-Subunit) Ab-2
Clone 17-69
Host Species Mouse
Quantity 100µL
Immunogen IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Isotype IgG1
Regulatory Status RUO
Conjugate Unlabeled
Research Discipline Neuroscience
Classification Monoclonal
Type Primary
Concentration 1mg/mL

Recommended for Immunoprecipitation, Thermo Scientific™ IGF-1 R (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.

Host Species: Mouse

Clone: 17-69

Isotype: IgG1

Species Reactivity: Human, Pig and Rabbit. Does not react with rat. Others not known.

Epitope: aa 514-586 (exon 7/8)

Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA

Molecular Weight: 125kDa

Positive Control: Placenta or Breast CA

Cellular Localization: Membrane Ab-2 is suitable for studies on pig and rabbit IGF-1R.

Recommended for:

  • Immunoprecipitation
  • Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
  • Does Not Significantly Inhibit IGF Binding

Note:

USA: RUO; Int'l: RUO