missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgE Rat anti-Mouse, Biotin, Clone: 23G3, eBioscience™

Rat Monoclonal Antibody

$117.48 - $236.28

Specifications

Antigen IgE
Clone 23G3
Host Species Rat
Gene Alias Immunoglobulin E, Ig E
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012108 Life Technologies
LS13599281
50μg Each for $117.48
5012109 Life Technologies
LS13599282
100μg Each for $236.28
Specifications

Description

The 23G3 monoclonal antibody reacts with the ε heavy chain of mouse IgE. It does not react with other classes of mouse immunoglobulin including IgD, IgG, IgA, or IgM.
Description & Specifications

Specifications

IgE
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
23G3
Immunoglobulin E, Ig E
ELISA,Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL