Please login to your online account to display your discounted pricing

IgE Mouse anti-Human, APC, Clone: Ige21, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen IgE
Clone Ige21
Host Species Murine
Gene Alias Immunoglobulin E
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123182

 
life technologies
LS17698641
25 Tests Each for $172.55

501123183

 
life technologies
LS17698642
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen IgE
Clone Ige21
Host Species Murine
Gene Alias Immunoglobulin E
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The monoclonal antibody IgE21 recognizes the immunoglobulin E class of antibodies. The natural levels of IgE in the serum are quite low, comprising only a small portion of the total immunoglobulin. IgE plays a role in allergy responses by binding to mast cells and basophils through the Fc epsilon receptor I which results in the release of histamine and other molecules. The IgE is produced by terminally differentiated plasma B cells and basophils. Like all immunoglobulins, IgE is expressed as a soluble molecule as well as a membrane bound form. IgE21 recognizes both forms.