missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgD Rat anti-Mouse, PerCP-eFluor 710, Clone: 11-26c (11-26), eBioscience™

Rat Monoclonal Antibody

$187.05 - $462.55

Specifications

Antigen IgD
Clone 11-26c (11-26)
Host Species Rat
Gene Alias Immunoglobulin D, Ig D
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123878 Life Technologies
LS46599382
100μg Each for $462.55
501128623 Life Technologies
LS46599380
25μg Each for $187.05
Specifications

Description

The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells.
Description & Specifications

Specifications

IgD
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
11-26c (11-26)
Immunoglobulin D, Ig D
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL